Imprimer
Fig tree Ficin, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0219876105 0219876125 0219876180
ICNA0219876105EA 83.4 EUR
ICNA0219876105 ICNA0219876125 ICNA0219876180
Fig tree Ficin, MP Biomedicals
Enzymes
  • EC 3.4.22.3
  • Refined ficin FSM200
  • Activity: >200 Bapa-U/gr hygroscopic