Imprimer
Aspergillus niger Type X glucose oxidase, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0219519690 0219519610 0219519650 0219519683
ICNA0219519690EA 938 EUR
ICNA0219519690 ICNA0219519610 ICNA0219519650 ICNA0219519683
Aspergillus niger Type X glucose oxidase, MP Biomedicals
Enzymes
  • Activity: >100 units/mg
  • Unit Definition: one unit oxidises 1,0 µmol of β-D-glucose to D-gluconic acid and H₂O₂ per minute at 35 °C, pH 5,1