Imprimer
Colistin sulphate, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0219415725 0219415783 0219415791
ICNA0219415725EA 56.2 EUR
ICNA0219415725 ICNA0219415783 ICNA0219415791
Colistin sulphate, MP Biomedicals
Protéines et Peptides
Colistin is a polymyxin antibiotic, and is a mixture of cyclic polypeptides colistin A and B. Colistin is effective against most Gram-negative bacilli and is used as a polypeptide antibiotic.
  • Storage temperature: +4 °C, desiccate
Mode of Action: Binds to lipids on the cell cytoplasmic membrane of Gram-negative bacteria and disrupts the cell wall integrity.
Antimicrobial spectrum: Gram-negative bacteria. It is proposed that renal reabsorption of colistin may involve organic cation transporters and peptide transporters and that the process is sensitive to pH.
Colistin sulphate is used to permeabilise bacterial cell membranes and to study mannose-resistant haemagglutination and antibiotic resistance in certain organisms, such as A. baumannii. It has been used to study hephrotoxicity in the rat kidney, and MICs, time-kill kinetics, and postantibiotic effect (PAE) against Pseudomonas aeruginosa.