Imprimer
Bovine lipoprotein-cholesterol concentrate (from Serum), MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0219147625 0219147680
ICNA0219147625EA 235 EUR
ICNA0219147625 ICNA0219147680
Bovine lipoprotein-cholesterol concentrate (from Serum), MP Biomedicals
Protéines et Peptides
Lipoprotein- cholesterol is a mixture of lipoprotein cholesterol, phospholipids and fatty acids.
  • Concentration: Cholesterol concentration: 1000 to 2000 mg/dl
  • Filtered through 0,2 µm membrane filter
For use as a serum-free or serum-reduced media nutrient supplement, calibration of cholesterol values, and enzymatic or non-enzymatic cholesterol determinations.