Imprimer

Levamisole hydrochloride, poudre cristalline blanche

Marques : MP Biomedicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

Synonyme(s): (-)-Tetramisole Hydrochloride, (-)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole Hydrochloride, (S)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride

0215522805 0215522810
ICNA0215522805EA 58.8 EUR
ICNA0215522805 ICNA0215522810
Levamisole hydrochloride, poudre cristalline blanche
Levamisole hydrochloride

Levamisole hydrochloride is suitable as an inhibitor of various mammalian alkaline phosphatases.


  • Intestinal alkaline phsophatases are only slightly inhibited
  • Biological response modifier with anthelmintic activity
  • Stereospecific inhibitor of succinic dehydrogenase.


Shows both immunostimulant and immunosuppressant effects, depending on several controllable factors. Very effective in treatment of ascariasis (hookworm infestation). Useful in chemotherapy of colorectal cancers, possibly due to its stimulation of IL-1 production and direct activation of macrophages.


Shows both immunostimulant and immunosuppressant effects, depending on several controllable factors. Very effective in treatment of ascariasis (hookworm infestation). Useful in chemotherapy of colorectal cancers, possibly due to its stimulation of IL-1 production and direct activation of macrophages.

Formule: C₁₁H₁₂N₂S·HCl
Poids moléculaire: 240.75 g/mol
Numéro MDL: MFCD00012675
Numéro CAS: 16595-80-5

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR