Imprimer

Recombinant IL2 (from E. coli), MP Biomedicals

Marques : MP Biomedicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

0215389610
ICNA0215389610EA 689 EUR
ICNA0215389610
Recombinant IL2 (from E. coli), MP Biomedicals
Protéines et Peptides

Interleukin-2 is an immunomodulatory factor produced by certain subsets of T lymphocytes. This lympho­kine promotes long term growth of activated T cells and related cell types.


  • Specific activity: ≥5×10⁶ µ/mg
  • Endotoxin: ≤0,15 ng/µ


It plays a role in the activation and proliferation of NK cells, induces γ-inter­feron and B cell growth factor secretion, and modulates the expression of the IL-2 receptor.

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR