Imprimer

3,7-Dihydro-3-méthyl-1H-purine-2,6-dione, poudre blanche

Marques : MP Biomedicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Attention

Synonyme(s): 3,7-Dihydro-3-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3-methylpurine

0215168680 0215168683
ICNA0215168680EA 32.5 EUR
ICNA0215168680 ICNA0215168683
3,7-Dihydro-3-méthyl-1H-purine-2,6-dione, poudre blanche
3,7-Dihydro-3-méthyl-1H-purine-2,6-dione

3-Methylxanthine, a metabolite of theophylline, was quantitated in rat plasma by a sensitive LC-MS/MS method.


Freely soluble in alkali hydroxides and in mineral acid; soluble in dilute aqueous base or ethanol; slightly soluble in hot water; practically insoluble in organic solvent

Formule: C₆H₆N₄O₂
Poids moléculaire: 166.14 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD00005580
Numéro CAS: 1076-22-8
EINECS: 214-058-1

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR