Imprimer

n-Dodecyl-β-D-maltoside ~97%, powder_white

Marques : MP Biomedicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): DDM

0215101380 0215101391
ICNA0215101380EA 38.8 EUR
ICNA0215101380 ICNA0215101391
n-Dodecyl-β-D-maltoside ~97%, powder_white
n-Dodecyl-β-D-maltoside

N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research.


It has also been used for the isolation of the mitochondrial cytochrome c oxidase.


Soluble in water (100 mg/mL) - clear to very slightly hazy, colorless solution.


Attention: Research Use Only (RUO).

Formule: C₂₄H₄₆O₁₁
Poids moléculaire: 510.6 g/mol
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00043012
Numéro CAS: 69227-93-6

Commander maintenant

Résultats des tests de spécification

Identity Test Passes
Purity ~97%
FTIR Conforms to Reference
Optical Rotation (c=1, methanol) +47 ° ± 5 °


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR