Imprimer
Pig Trypsin (from Pancreas), MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
0210313980 0210313991 0210313920 0210313925 0210313983
ICNA0210313980EA 397 EUR
ICNA0210313980 ICNA0210313991 ICNA0210313920 ICNA0210313925 ICNA0210313983
Pig Trypsin (from Pancreas), MP Biomedicals
Enzymes
  • Dry powder: stable
  • Protease act: min 250,000 USP units/gm.
pH: ~ 8,0 (Optimum) (Lit.)