Imprimer

L(-)-Proline ≥99%, poudre blanche

Marques : MP Biomedicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): (S)-(-)-Proline, H-Pro-OH

0210273025 0210273080 0210273091
ICNA0210273025EA 34.5 EUR
ICNA0210273025 ICNA0210273080 ICNA0210273091
L(-)-Proline ≥99%, poudre blanche
L(-)-Proline

L-Proline is an amino acid. The conjugation of L-proline with imidazolium-based ionic liquid (IL)-modified magnetic nanoparticles forms a catalyst that can be magnetically recovered and employed in direct asymmetric aldol reaction. The solubility and stability of ezetimibe (EZT) has been reported to enhance on cocrystallization with L-proline.

L-Proline has been used:


  • As a supplement during the preparation of chondrogenic medium and synthetic dextrose minimal medium (SD).
  • As a standard during the identification of metabolites in serum samples.
  • In a study to prepare L-proline-L-phenylalanine (L-Pro-L-Phe) mixture in aqueous acetonitrile.


L-Proline is used in cell culture media and is a component of MEM non-essential amino acids solution.In cell culture research, L-proline has been used in studies of MOLT-4 cells in starvation-induced apoptosis, maintenance of hematopoietic cell cultures derived from the prawn nephrops norvegicus, growth behavior of various Escherichia coli strains at various L-proline concentrations, and rat primary hepatocytes in a collagen gel sandwich culture.


Soluble in alcohol and water (40 mg/ml-clear, colorless solution); insoluble in ether, butanol isopropanol.


Attention: Research Use Only (RUO).

Formule: C₅H₉NO₂
Poids moléculaire: 115.13 g/mol
Point d'ébullition: 245
Point de fusion: 221 °C (déc.)
Température de stockage: Température ambiante
Numéro MDL: MFCD00064318
Numéro CAS: 147-85-3
Indice Merck: 13,07871

Commander maintenant

Résultats des tests de spécification

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
Loss on Drying ≤0.4%
Optical Rotation (c=1.6, water) -82° to -85°
Hydroxy-L-proline Free


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR