Imprimer

(±)-Acide 5-(dithiolanne-3-yl)valerique ≥98%, en poudre

Marques : MP Biomedicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): (±)-α- Acide Lipoique, DL-Acide Thioctique (oxydé)

0210113825 0210113801 0210113805 0210113850
ICNA0210113825EA 299 EUR
ICNA0210113825 ICNA0210113801 ICNA0210113805 ICNA0210113850
(±)-Acide 5-(dithiolanne-3-yl)valerique ≥98%, en poudre
(±)-Acide 5-(dithiolanne-3-yl)valerique

DL-Thioctic Acid is a hydrogen transferrin cofactor.


Antioxidant and coenzyme needed for the activity of enzyme complexes such as those of pyruvate dehydrogenase and glycine decarboxylase. Exogenous thioctic acid is reduced intracellularly by two or more enzymes. The reduced form then influences a number of cell processes by direct radical scavenging, recycling of other antioxidants, increasing glutathione synthesis, and modulating transcription factor activity particularly that of NF-κB. Decreases the phagocytosis of myelin by macrophages.


Soluble in fat solvents, methanol (50 mg/ml - clear pale yellow solution), ethanol(50 mg/ml - clear solution); practically insoluble in water.

Formule: C₈H₁₄O₂S₂
Poids moléculaire: 206.33 g/mol
Numéro MDL: MFCD00005474
Numéro CAS: 1077-28-7
EINECS: 214-071-2

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR