Imprimer
Tank liners, HyPerforma™
Fournisseur: THERMO BIOPROCESS DIVISION
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
SH30647.04 SH30647.01 SH30647.05 SH30647.02 SH30647.03 SH30646.01 SH30646.02 SH30646.03
HYCPSH30647.04EA 1960 EUR
HYCPSH30647.04 HYCPSH30647.01 HYCPSH30647.05 HYCPSH30647.02 HYCPSH30647.03 HYCPSH30646.01 HYCPSH30646.02 HYCPSH30646.03
Tank liners, HyPerforma™
Sachets Sachets pour milieux de culture
Tank liners are gamma irradiated, open single use bags designed to fit in cylindrical drums.
  • Gusset design allows for a close fit to the tank
  • Constructed of CX3-9 film
  • For HyPerforma™ Single-Use Mixer DS 300