Imprimer
Circulating cooling thermostats, unichillers, with Pilot ONE E-grade "Basic" controller
Fournisseur: HUBER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
3012.0189.01 3038.0001.01 3038.0010.01 3038.0003.01 3038.0011.01 3038.0027.01 3038.0028.01 3038.0004.01 3038.0016.01 3038.0005.01 3038.0013.01 3038.0029.01 3038.0030.01 3040.0009.01 3040.0010.01 3038.0043.01 3038.0044.01 3040.0011.01 3040.0012.01 3038.0045.01 3038.0046.01
HUBE3012.0189.01EA 0 EUR
HUBE3012.0189.01 HUBE3038.0001.01 HUBE3038.0010.01 HUBE3038.0003.01 HUBE3038.0011.01 HUBE3038.0027.01 HUBE3038.0028.01 HUBE3038.0004.01 HUBE3038.0016.01 HUBE3038.0005.01 HUBE3038.0013.01 HUBE3038.0029.01 HUBE3038.0030.01 HUBE3040.0009.01 HUBE3040.0010.01 HUBE3038.0043.01 HUBE3038.0044.01 HUBE3040.0011.01 HUBE3040.0012.01 HUBE3038.0045.01 HUBE3038.0046.01
Circulating cooling thermostats, unichillers, with Pilot ONE E-grade "Basic" controller
Thermostats à circulation Systèmes à circulation fermée Circulateurs de refroidissement
Unichillers offer better efficiency, stable pressure and delivery performance with a constant cooling water temperature compared to traditional tap water cooling. Chillers also reduce water consumption and help conserve the environment whilst lowering operating costs.
  • Simple operation with menu navigation
  • Robust stainless construction
  • Easy to fill and empty
  • Illuminated filling level indicator
  • Bright, large OLED display
Unichillers are ideal for removing heat from chemical processes, cooling technical plants or as a central cooling water supply forEn savoir plus