Imprimer
Interfaces
Fournisseur: HUBER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
6915 31217 10019
HUBE6915EA 873 EUR
HUBE6915 HUBE31217 HUBE10019
Interfaces
Prises électriques
Com.G@te®. The interfaces are integrated RS232 (bi-directional), RS485 (bi-directional), ECS external control signal, volt free contact, AIF analogue interface 0/4 to 20 mA or 0 to 10 V (bi-directional). Connections comply with NAMUR standard.