Imprimer

Silver methenamine

Marques : SIGMA ALDRICH MICROSCOPY
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger


MQ500

HT1003-6VL
HT1003-6VLEA 308 EUR
HT1003-6VL
Silver methenamine
Silver methenamine

Intended for use in the silver stain (modified GMS) procedure.

Formule: C₆H₁₂AgN₄
Poids moléculaire: 248.06 g/mol
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00284609
Numéro CAS: 37604-90-3

Commander maintenant

Résultats des tests de spécification

Appearance (Colour) White to Light Grey
Appearance (Form) Powder
Performance Conforms
24 months When using Sigma-Aldrich Procedure No. HT100, fungi organisms stain purplish-brown to black


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR