Imprimer
Ice Maker, HC Series
Fournisseur: HOSHIZAKI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
FM-120KE-HC FM-120KE-HCN
HOSHFM-120KE-HCEA 5845 EUR
HOSHFM-120KE-HC HOSHFM-120KE-HCN
Ice Maker, HC Series
Machines à glace
The FM-120KE-HC/HCN are self-contained ice makers, which utilise natural refrigerant R290.
  • Automatic flush system to reduce sediment build up
  • Stainless steel exterior with integrated door handles provides a hygienic and high quality finish
  • Member of the Emerald class,Hoshizaki’s range of climate friendly ice makers
  • Quality of ice is always consistent
Closed water circuit offers the ultimate contamination protection,by reducing the number of points at which impurities can enter the ice making process.
Electronically controlled to ensure an optimisedEn savoir plus