Imprimer
Thallium AAS Standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
E4078-100ML E4078-500ML
HONCE4078-100MLEA 50.9 EUR
HONCE4078-100ML HONCE4078-500ML
Thallium AAS Standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de thallium
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).