Imprimer
Sodium AAS Standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
E4048-100ML E4048-500ML E4049-100ML E4049-500ML E4050-100ML E4050-500ML E4112-100ML E4204-100ML E4204-500ML
HONCE4048-100MLEA 44.3 EUR
HONCE4048-100ML HONCE4048-500ML HONCE4049-100ML HONCE4049-500ML HONCE4050-100ML HONCE4050-500ML HONCE4112-100ML HONCE4204-100ML HONCE4204-500ML
Sodium AAS Standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de sodium
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).