Imprimer
Sodium ICP Standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
E1037-100ML E1113-100ML E1164-100ML E1233-100ML
HONCE1037-100MLEA 101 EUR
HONCE1037-100ML HONCE1113-100ML HONCE1164-100ML HONCE1233-100ML
Sodium ICP Standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de sodium
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).