Imprimer
Manganese ICP standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
E1035-100ML E1111-100ML E1162-100ML
HONCE1035-100MLEA 101 EUR
HONCE1035-100ML HONCE1111-100ML HONCE1162-100ML
Manganese ICP standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de manganèse
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).