Imprimer
Potassium ICP Standards, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
E1030-100ML E1108-100ML E1157-100ML E1227-100ML
HONCE1030-100MLEA 102 EUR
HONCE1030-100ML HONCE1108-100ML HONCE1157-100ML HONCE1227-100ML
Potassium ICP Standards, (CRMs), Fluka™
Étalons Étalons cationiques Standards de potassium
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).