Imprimer
Iridium ICP Standard, (CRMs), Fluka™
Marques : Honeywell Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
E1029-100ML
HONCE1029-100MLEA 372 EUR
HONCE1029-100ML
Iridium ICP Standard, (CRMs), Fluka™
Étalons Étalons cationiques Standards d'iridium
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable