Imprimer
UV-Vis Spectrophotometers, U-3900S and U-3900D
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
2J2-0063 2J2-0073
HITA2J2-0063EA 17750 EUR
HITA2J2-0063 HITA2J2-0073
UV-Vis Spectrophotometers, U-3900S and U-3900D
Spectrophotomètres et Photomètres Spectrophotomètres UV-Visible
UH3900S (Single monochromator) and UH3900D (Double monochromator) are reliable and proven optical systems equipped with new Software UV Solutions Plus.
  • Extensive applications
  • Full range of accessories for liquid and solid samples
  • Ease of maintenance (lamp replacement)
  • Stable baseline
  • Effective in trace sample measurement
U-3900S and U-3900D spectrophotometers are comprehensive, research-grade solutions. They provide excellent performance afforded by low stray light, selectable band pass and high energy. Hitachi Model UH3900S/D spectrophotometer adopts
En savoir plus