Imprimer
Pipette stand, rotary
Fournisseur: HIRSCHMANN
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
8902101
HIRS8902101EA 108 EUR
HIRS8902101
Pipette stand, rotary
Portoirs pour pipettes
Pipette stand made from blue PP.
  • Rotatable model with a reinforced base and an additional, rotatable disk to protect the pipette tips
  • 94 holes with different diameters for graduated and volumetric pipettes of any size