Imprimer
Avantor® Alltima HP, HPLC columns, HILIC, 3 µm
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
HICH86461EA 731 EUR
HICH86461 HICH86463 HICH86462
Avantor® Alltima HP, HPLC columns, HILIC, 3 µm
Colonnes pour chromatographie
Avantor® Alltima HP columns features hydrophilic interaction chromatography that uses small amounts of water for increased sensitivity
with microbore applications.
  • High resolution for complex samples
  • High purity silica
  • Excellent column stability
These columns combine the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.
These are widely used for polar analytes that are difficult to retain by reversed-phase.