Imprimer
Networking kit with Centri-Log™ data collection software
Fournisseur: Thermo Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
75007602 75007516 75007517 75007535 75007702
HERA75007602EA 2960 EUR
HERA75007602 HERA75007516 HERA75007517 HERA75007535 HERA75007702
Networking kit with Centri-Log™ data collection software
Logiciels Logiciels de laboratoire
Centri-Log™ data collection software is a paperless process tracking and compliance solution. This innovative software provides effective quality control for the documentation (logging and monitoring) of centrifugation processes, enabling lifecycle management of processed samples, equipment optimisation, and compliance with standard operating procedures.
  • Up to 30 centrifuges can be integrated in a single network and remotely managed from a single desktop
  • Quick, easy, secure and accurate data collection and document management
  • Continual monitoring of speed,
En savoir plus