Imprimer
Hei-VAP Ultimate Rotary Evaporators
Fournisseur: Heidolph Instruments GmbH & Co.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
575-01100-00 575-01300-00 575-01305-00 575-01500-00 575-01600-00 575-01110-00 575-01310-00 575-01315-00 575-01510-00 575-01610-00 576-01100-00 576-01300-00 576-01305-00 576-01500-00 576-01600-00 576-01110-00 576-01310-00 576-01315-00 576-01510-00 576-01610-00
HEID575-01100-00EA 5520 EUR
HEID575-01100-00 HEID575-01300-00 HEID575-01305-00 HEID575-01500-00 HEID575-01600-00 HEID575-01110-00 HEID575-01310-00 HEID575-01315-00 HEID575-01510-00 HEID575-01610-00 HEID576-01100-00 HEID576-01300-00 HEID576-01305-00 HEID576-01500-00 HEID576-01600-00 HEID576-01110-00 HEID576-01310-00 HEID576-01315-00 HEID576-01510-00 HEID576-01610-00
Hei-VAP Ultimate Rotary Evaporators
Évaporateurs Évaporateurs rotatifs
The Hei-VAP ultimate, precise setting of rotation speed and heating bath temperature on the digital touch display or for quick access during an active process directly via the knobs on the operating panel.
  • Central operating panel with 7" touch display
  • Clear display of all values (basic and advanced modes)
  • With timer, limitation of max. temperature, multilingual menu and text display for error messages
  • Separate knobs for rotation and heating with locking function to prevent accidental readjusting
  • LED ring lights and pictogram to display the activity even
En savoir plus