Imprimer
Watch glasses
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
40010020 40010011 92001200020 40010004 40010005 40010006 40010007 40010008 40010009 40010010 40010012 40010014 40010015 40010018
HECH40010020EA 116 EUR
HECH40010020 HECH40010011 HECH92001200020 HECH40010004 HECH40010005 HECH40010006 HECH40010007 HECH40010008 HECH40010009 HECH40010010 HECH40010012 HECH40010014 HECH40010015 HECH40010018
Watch glasses
Verres de montre
Evaporating dishes, made of glass, with fused rim.
  • Good chemical restistance
  • Available with or without hole
Certifications: DIN 12 341