Imprimer
Counting chambers Sedgewick Rafter
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
40430010
HECH40430010EA 1270 EUR
HECH40430010
Counting chambers Sedgewick Rafter
Lames Cellules de numération
Sedgewick rafter counting chambers are used for counting particles and microorganisms in 1 ml of water or other transparent liquids, among other things for drinking water analysis. The counting chambers are made of high-quality optical glass with a chrome grid (counting net). The cell = 1 cm³ is graduated with a 1mm grid that divides 1 ml into 1000 μl.
Glass counting chamber with chrome grid for phase contrast with ground and faceted edges, including cover glass.