Imprimer
Cover glasses for hemacytometers, Assistent®
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
40415710 40416710 40412710 40417710
HECH40415710EA 36.4 EUR
HECH40415710 HECH40416710 HECH40412710 HECH40417710
Cover glasses for hemacytometers, Assistent®
Lamelles couvre-objet
Assistent® counting chambers are precision items in special optical glass used to count the particles per volume unit of a liquid.
  • Lateral fields for inscription separated by grooves from the three ground and polished supports
  • Central support deeper than the outer ones and referred to as depth of chamber
  • Central support of those chambers divided by an additional groove, coated by a rhodium layer
Certifications: IVD directive 98/79/EC