Imprimer
Pipette workstation
Fournisseur: HEATHROW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
HS234631
HEATHS234631EA 292 EUR
HEATHS234631
Pipette workstation
Portoirs pour pipettes
Keep pipette and disposable tips together in one convenient stand.
  • Holds up to four pipettes on top shelf
  • Dual bins hold multiple size tips
  • Self-closing lid provides dust-free storage
  • Oversized lip allows for easy access when wearing gloves
Acrylic workstation is opaque on sides with clear front and bin lids for easy content identification.