Imprimer
Slide staining tray
Fournisseur: HEATHROW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
HS15951A
HEATHS15951AEA 192 EUR
HEATHS15951A
Slide staining tray
Boîtes de coloration
Made of a PP and PE blend, this rugged staining tray is a good alternative to metal version. Slide staining tray set, disposable. Provided with four base trays and one lid.
  • Stain, rinse, and dry slides on a single working tray
  • A clean and fresh working surface area every time
  • Compact size and recessed handles for easy transport from work area to sink
  • Two convenient pour spouts for quick disposal of unwanted liquid waste
  • Dark lid protects slides for light sensitive applications
This disposable slide staining tray set reduces possible crossEn savoir plus