Imprimer
Knife spatulas, angled
Fournisseur: HAMMACHER KARL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
HSN236-10 HSN237-13
HAMMHSN236-10EA 114 EUR
HAMMHSN236-10 HAMMHSN237-13
Knife spatulas, angled
Spatules
Flexible knife spatulas with wooden handle and blade made of stainless steel.