Imprimer

N-Formyl-L-methionine 95%

Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): For-Met-OH, Formyl-L-methionine

H63493.MD H63493.03 H63493.06
H63493.MDEA 38.2 EUR
H63493.MD H63493.03 H63493.06
N-Formyl-L-methionine 95%
N-Formyl-L-methionine

Purity: 95% CAS No.: 4289-98-9 Molecular Formula: C6H11NO3S Formula Weight: 177.22 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD00021033

Formule: C₆H₁₁NO₃S
Poids moléculaire: 177.2 g/mol
Numéro MDL: MFCD00021033
Numéro CAS: 4289-98-9

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR