Imprimer
VACUETTE® Blood Collection Tubes, without Additive
Fournisseur: Greiner Bio-One
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
456085_1200
GREI456085_1200EA 202 EUR
GREI456085_1200
VACUETTE® Blood Collection Tubes, without Additive
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons de sang
Blood collection tubes made from practically unbreakable plastic
These tubes do not contain any additives and are used for CSF determinations or puncture specimens. Without threads, the tubes are especially suitable for automatic decapping by a quick pull movement.
They have made the drawing of blood a safer and more hygienic process. The use of PET plastic offers the advantage that the tubes are as clear as glass.