Imprimer
CO₂ Resistant Orbital Shaker, Grant Instruments
Fournisseur: GRANT INSTRUMENTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
CPS-20
GRANCPS-20EA 2450 EUR
GRANCPS-20
CO₂ Resistant Orbital Shaker, Grant Instruments
Agitateurs Agitateurs orbitaux
The CPS-20 is a CO₂ resistant shaker providing regulated orbital motion and designed for use specifically in CO₂ incubators.
  • Designed for use in harsh environments such as CO₂ and humidity
  • Choice of five interchangeable platforms available enabling a wide variety of vessels to be used (must be ordered separately)
  • Specifically designed remote controller provides protection for the electronics from the CO₂ environment
  • Brushless motor with a guaranteed life time of 35000 hours
The CPS-20 is specifically designed for use in harsh environments such as CO₂
En savoir plus