Imprimer
eStain™ L1 Protein Staining Device
Fournisseur: Genscript
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
L00658 L00660 L00661-10 L00662-10 L00663
GENSL00658EA 603 EUR
GENSL00658 GENSL00660 GENSL00661-10 GENSL00662-10 GENSL00663
eStain™ L1 Protein Staining Device
Boîtes de coloration
The eStain™ is a fully automated protein PAGE gel staining system, which uses Coomassie Brilliant Blue and a patented protein staining technology. The system integrates the traditional three steps of fixing, staining, and destaining into one automated process. It can stain and destain two protein PAGE gels simultaneously in 10 minutes.
  • Automated 10-minute staining and destaining for 2 gels simultaneously
  • Consistent results with coomassie blue staining (offered by GenScript) or membrane staining with Ponceau S solution
  • Easy to use with clean setup and
En savoir plus