Imprimer

Anti-SUMO-tag Mouse Monoclonal Antibody [clone: 4G11E9]

Fournisseur: Genscript
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

A01693-40
GENSA01693-40EA 173 EUR
GENSA01693-40
Anti-SUMO-tag Mouse Monoclonal Antibody [clone: 4G11E9]
Anticorps
Type: Tag
Antigen: SUMO-tag
Clonality: monoclonal
Clone: 4G11E9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR