Imprimer

Anti-MYC Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: Genscript
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

A00863-100
GENSA00863-100EA 311 EUR
GENSA00863-100
Anti-MYC Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Type: Tag
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR