Imprimer
Magnetic stand
Fournisseur: G-Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
786-888
GENO786-888EA 661 EUR
GENO786-888
Magnetic stand
Supports de statif
Magnetic stand is designed for the efficent pull down of magnetic beads in 1,5 and 2 ml micro centrifgue tubes.
  • Suitable for 8×1,5 to 2 ml tubes
  • Pulls beads to tube wall to allow removal of all supernatant
The powerful magnet pulls beads to the side of the tube, allowing for the removal of all the liquid supernatant with no contamination with beads. It can also be used for immunoprecipitations and protein purification analysis.