Imprimer

Eau, Distilled, sterile

Marques : G Biosciences
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): Hydrogen oxide

786-1712 786-1711 786-1713 786-1715 786-1714 786-1716 786-1717 786-1718
GENO786-1712EA 38.5 EUR
GENO786-1712 GENO786-1711 GENO786-1713 GENO786-1715 GENO786-1714 GENO786-1716 GENO786-1717 GENO786-1718
Eau, Distilled, sterile
Eau

Laboratory reagent grade, sterile distilled water for your laboratory needs. This water is purified by steam distillation and sterile filtered for use as a research laboratory reagent. The distilled water is low endotoxin <0.025EU/ml and low conductivity (<10uS). This water is ideal for the preparation of reagents, rinsing of glassware and plasticware, and various other laboratory and research applications. No toxic agents such as DEPC are included in the preparation of distilled water, eliminating interference with enzymatic analysis.

Formule: H₂O
Poids moléculaire: 18.02 g/mol
Point d'ébullition: 100
Point de fusion: 0
Numéro MDL: MFCD00011332
Numéro CAS: 7732-18-5
Indice Merck: 13,10098

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR