Imprimer

Human Serum Albumin (HSA) Solution (20%), 50 ml

Fournisseur: GEMINI BIO-PRODUCTS
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

800-121-050
GEMB800-121-050EA 183 EUR
GEMB800-121-050
Human Serum Albumin (HSA) Solution (20%), 50 ml
Sérums
This diagnostic-grade product is an aqueous solution containing 20% human albumin (w/v). It is prepared by cold-alcohol fractionation from pooled human plasma obtained from venous blood. The USP Pharmacopeia requires that all fractionators add sodium caprylate and sodium tryptophanate as a preservative.

Attention: For cell culture, further manufacturing and research use only. Not for direct therapeutic use.
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR