Imprimer

Glucose assay reagent, Supelco®

Marques : Merck
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

G3293-50ML G3293-20ML
G3293-50MLEA 0 EUR
G3293-50ML G3293-20ML
Glucose assay reagent, Supelco®
Tests pour aliments et boissons

Activity <0,05% resulting from cross-reactivity with the following substrates: fructose, dextran, sedoheptulose, arabinose, galactose, xylose, trehalose, mannose, sorbitol, fucose or ribose.


For the quantitative determination of glucose in food and other materials utilizing the hexokinase-glucose 6-phosphate dehydrogenase enzymatic assay. Reconstitute with deionised water using volume indicated by package size (20 or 50 ml).


Cdt: 20, 50 ml in serum bottle.

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR