Imprimer
Cohan-Vannas spring scissors
Fournisseur: FINE SCIENCE TOOLS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
15000-11 15000-02 15000-01 15000-12
FSCI15000-11EA 745 EUR
FSCI15000-11 FSCI15000-02 FSCI15000-01 FSCI15000-12
Cohan-Vannas spring scissors
Ciseaux
These stainless steel scissors have long cutting blades for a more controlled cut.