Imprimer
Vacuum wand, C-series
Fournisseur: WINDRUSH TECHNOLOGY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
106518 90-CP C002-Y-92-CP C002-Y-90-CP C003-X-95-CP 98-CP C003-Y-99-CP C003-Y-91-CP C002-X-95-CP C003-Y-92-CP C002-Y-98-CP C003-X-96-CP C001-Y-91-CP C002-X-96-CP 99-CP 91-CP C002-Y C003-Y-90-CP
FLUM106518EA 0 EUR
FLUM106518 FLUM90-CP FLUMC002-Y-92-CP FLUMC002-Y-90-CP FLUMC003-X-95-CP FLUM98-CP FLUMC003-Y-99-CP FLUMC003-Y-91-CP FLUMC002-X-95-CP FLUMC003-Y-92-CP FLUMC002-Y-98-CP FLUMC003-X-96-CP FLUMC001-Y-91-CP FLUMC002-X-96-CP FLUM99-CP FLUM91-CP FLUMC002-Y FLUMC003-Y-90-CP
Vacuum wand, C-series
Systèmes de manipulation sous vide
This C series wand bodies are manufactured in ESD safe materials including PEEK tips are the safest means of wafer handling. The tips are optimally sized for each wafer diameter, ensuring that contact is both minimal and secure.
  • Polished contact surface provides excellent adhesion to water
  • Fixed tip and swivel joint body styles are available
  • Unique control valve ensures reliable suction and instantaneous release without particle generation