Imprimer
Exsil C8, HPLC Columns
Fournisseur: Avantor
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
EXC8-3-150AEA 439 EUR
EXC8-3-150A
Exsil C8, HPLC Columns
Colonnes pour chromatographie
Exsil HPLC columns are a useful alternative to the classical spherical HPLC silicas.
  • Spherical porous silica
  • Competitively priced
  • High efficiency
Exsil 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications. Exsil 80 is a higher surface area, complementary range of materials.