Imprimer
Conductivity meter, bench top, Eutech CON 2700
Fournisseur: EUTECH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
ECCON270043S
EUTEECCON270043SEA 1170 EUR
EUTEECCON270043S
Conductivity meter, bench top, Eutech CON 2700
Compteurs électrochimiques Conductimètres
The Eutech CON 2700 accepts 2-cell and 4-cell electrodes, allowing a broad range of measurement at up to ±1 % full scale accuracy. It can measure conductivity, TDS, salinity, resistivity and the temperature.
  • Multi data screen shows conductivity readings, temperature, electrode status, calibration points, date and time
  • Auto-calibration with preset values for quick easy calibration, or manual multi-point calibration for greater accuracy
  • Quick, easy electrode diagnosis with the effective cell constants display
  • Non-volatile memory holds up to 500 data points –
En savoir plus