Imprimer
Abbe refractometer
Fournisseur: EUROMEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
98.490
EUMX98.490EA 1800 EUR
EUMX98.490
Abbe refractometer
Réfractomètres Réfractomètres de paillasse
The Abbe laboratory refractometer is a bench-top instrument for high-precision measurements of an index of refraction.
  • More accurate than hand refractometers
  • Capable of measuring all kinds of concentrations and identifying several types of substances
It can be connected to a water bath for measuring at a controlled temperature and used for determining the refractive index of glass, plastic films. During operating this type of refractometer, the use of a cold light source is recommended. The instrument is equipped with a built-in thermometer and waterEn savoir plus