Imprimer
Laboratory fume hoods, low flow rate
Fournisseur: EQUIP LABO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
55ZZ 57ZZ 45ZZ 54ZZ 53ZZ 52ZZ 51ZZ 49ZZ 48ZZ 46ZZ 44ZZ 43ZZ 42ZZ 47ZZ 56ZZ 67ZZ 78ZZ 77ZZ 76ZZ 75ZZ 74ZZ 73ZZ 70ZZ 68ZZ 82ZZ 66ZZ 65ZZ 64ZZ 63ZZ 62ZZ 59ZZ 58ZZ 69ZZ 79ZZ
EQUI55ZZEA 0 EUR
EQUI55ZZ EQUI57ZZ EQUI45ZZ EQUI54ZZ EQUI53ZZ EQUI52ZZ EQUI51ZZ EQUI49ZZ EQUI48ZZ EQUI46ZZ EQUI44ZZ EQUI43ZZ EQUI42ZZ EQUI47ZZ EQUI56ZZ EQUI67ZZ EQUI78ZZ EQUI77ZZ EQUI76ZZ EQUI75ZZ EQUI74ZZ EQUI73ZZ EQUI70ZZ EQUI68ZZ EQUI82ZZ EQUI66ZZ EQUI65ZZ EQUI64ZZ EQUI63ZZ EQUI62ZZ EQUI59ZZ EQUI58ZZ EQUI69ZZ EQUI79ZZ
Laboratory fume hoods, low flow rate
Hottes et coffrets Hottes filtrantes
Fume hoods which are specially designed for labs with low flow rates are of three types they are registre, caisson and attaque.
Registre: It is highly desigend to handle all types of solvents and volatiles, it is also indicated for the treatment of powders, fumes and steams.
Caisson: It is highly designed for acids, bases, heavy vapors and products requiring condensate recovery.
Attaque: It is designed for acids, acid heated and corrosive products requiring condensate recovery. Equipped with a washing box.