Imprimer
Soil sampler
Fournisseur: EP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
TERRACORE5G
EPSCTERRACORE5GEA 1080 EUR
EPSCTERRACORE5G
Soil sampler
Échantillonneurs Préleveurs de solides
This rugged all-in-one item eliminates the need for a separate handle.
  • More convenient than make-your-own cut-off syringe
  • Simple operation for easy and effective sampling
  • Minimizes handling and reduces target analyte loss
Designed for soil sampling into a 40 ml VOA vial for in-field preservation.